missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00078987-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MAPWPPKGLVPAVLWGLSLFLNLPGPIWLQPSPPPQSSPPPQPHPCHTCRGLVDSFNKGLERTIRDNFGGGNTAWEEENLSKYKDSETRLVEVLEGVCSKSDFECHRLLELSEELVESWWFHKQQEAPDLFQWLCSDSLKLCCPAGTFGPSCLPCPGGTERPCGGYGQCEGEGTRGGSGHCDCQAGYGGEACGQCGLGYFEAERNASHLVCSACFGPCARCSGPEESNCLQCKKGWALHHLKCVDIDECGTEGANCGADQFCVNTEGSYECRDCAKACLGCMGAGPGRCKKCSPGYQQVGSKCLDVDECETEVCPGENKQCENTEGGYRCICAEGYKQMEGICVKEQIPGAFPILTDLTPETTRRWKLGSHPHSTYVKMKMQRDEATFPGLYGKQVAKLGSQSRQSDRGTRLIHSQQASSQRSpecifications
| CRELD1 | |
| Polyclonal | |
| No additive | |
| NM_001031717.1 | |
| CRELD1 | |
| CRELD1 (NP_001026887.1, 1 a.a. ∼ 422 a.a) full-length human protein. | |
| RUO | |
| 78987 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| cysteine-rich with EGF-like domains 1 | |
| AVSD2/CIRRIN/DKFZp566D213 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction