missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRELD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112344
This item is not returnable.
View return policy
Description
This product can be used in direct ELISA.
CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
Specifications
| CRELD2 | |
| Polyclonal | |
| Unconjugated | |
| CRELD2 | |
| 5730592L21Rik; C85758; CREL2; CRELD2; cysteine rich with EGF like domains 2; cysteine rich with EGF-like domains 2; cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2; RGD1310614; UNQ185/PRO211 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 76737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9CYA0 | |
| CRELD2 | |
| Recombinant Mouse CRELD2 Protein. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction