missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRF Polyclonal Antibody
GREENER_CHOICE

Product Code. 17209033 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17209033 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17209033 Supplier Invitrogen™ Supplier No. PA5116529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, Mouse liver, Mouse brain, Rat liver, Rat brain Immunogen sequence: LLSRGPVPGA RQAPQHPQPL DFFQPPPQSE QPQQPQARPV LLRMGEEYFL RLGNLNKSPA APLSPASSLL AGGSGSRPSP EQATANFFRV LLQQLLLPRR SLDSPAALAE RGARNALGGH QEAPERERRS EEPPISLDLT FHLLREVLEM ARAEQLAQQA HSNRKLMEII.

Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRH
Gene Accession No. P01143, P06850, Q8CIT0
Gene Alias Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF
Gene Symbols CRH
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12918, 1392, 81648
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.