missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114486
This item is not returnable.
View return policy
Description
Positive controls that may be used include: Human Placenta Tissue Lysate, Human Brain Tissue Slide.
Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
Specifications
| CRF | |
| Polyclonal | |
| Unconjugated | |
| CRH | |
| Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P06850 | |
| CRH | |
| CRH antibody was raised against a 16 amino acid peptide near the amino terminus of human CRH. The immunogen is located within amino acids 30 - 80 of CRH. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction