Learn More
Tocris Bioscience™ CRID3 sodium salt
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CRID3 sodium salt is a potent NLRP3 inflammasome inhibitor; closes the active conformation of NLRP3 to the inactive state. Directly interacts with the Walker B motif within the NLRP3 NACHT domain. Inhibits IL-1β, IL-18 and IL-1α production (IC50 values are 7.2, 10.3 and 12-18 nM, respectively). Selective for NLRP3 over NLRC4 inflammasome and Toll-like receptor signaling. Reduces severity of experimental autoimmune encephalomyelitis, skin inflammation and airway inflammation in mice. Also glutathione S-transferase omega 1 inhibitor. Orally bioavailable.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | CRID3 sodium salt |
| Purity | 98% |
| Molecular Weight (g/mol) | 426.46 |