missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIM1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81803JF646
This item is not returnable.
View return policy
Description
CRIM1 Polyclonal antibody specifically detects CRIM1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CRIM1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 51232 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Janelia Fluor 646 | |
| CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52 | |
| CRIM1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human Crim1. The immunogen is located within amino acids 910 - 960 of CRIM1. Amino Acid Squence: DYRDNRLHPSEDSSLDS | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction