missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597618
This item is not returnable.
View return policy
Description
CRIP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CRIP1 is a cysteine-rich intestinal protein (CRIP) that belongs to the LIM/double zinc finger protein family, members of which include cysteine- and glycine-rich protein-1 (CSRP1 MIM 123876), rhombotin-1 (RBTN1 MIM 186921), rhombotin-2 (RBTN2 MIM 180385), and rhombotin-3 (RBTN3 MIM 180386). CRIP may be involved in intestinal zinc transport.
Specifications
| CRIP1 | |
| Polyclonal | |
| Unconjugated | |
| CRIP1 | |
| CRHP; Crip; CRIP1; CRP1; CRP-1; cysteine rich intestinal protein; cysteine rich protein 1; cysteine-rich heart protein; Cysteine-rich intestinal protein; cysteine-rich protein 1; cysteine-rich protein 1 (intestinal) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1396 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50238 | |
| CRIP1 | |
| Recombinant Human Cysteine-rich protein 1 protein (1-77AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction