missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIP2 Antibody (05), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06566APC
This item is not returnable.
View return policy
Description
CRIP2 Monoclonal antibody specifically detects CRIP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CRIP2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1397 | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 05 | |
| Immunohistochemistry-Paraffin | |
| CRIP, CRP-2, CRP2ESP1, cysteine-rich protein 2, Cystein-rich intestinal protein, Protein ESP1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CRIP2 (Uniprot#: P52943; Met1-Pro208). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction