missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIP2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18431AF532
This item is not returnable.
View return policy
Description
CRIP2 Polyclonal antibody specifically detects CRIP2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CRIP2 | |
| Polyclonal | |
| Western Blot | |
| CRIP, CRP-2, CRP2ESP1, cysteine-rich protein 2, Cystein-rich intestinal protein, Protein ESP1 | |
| Recognizes region between residues Between Between 158 and 208.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1397 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction