missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIPTO Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1598PCP
This item is not returnable.
View return policy
Description
CRIPTO Polyclonal antibody specifically detects CRIPTO in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| CRIPTO | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| PerCP | |
| CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1 | |
| A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50). [UniProt# P51865] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Neuroscience, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 6997 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction