missing translation for 'onlineSavingsMsg'
Learn More

Crisaborole, MedChemExpress

Product Code. 16425246
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16425246

Brand: Medchem Express HY10978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Crisaborole (AN-2728) is a potent inhibitor of PDE4 and cytokine release; inhibit PDE4 with an IC50 of 0.49 μM.

Crisaborole (AN-2728) is a potent inhibitor of PDE4 and cytokine release

Specifications

Chemical Name or Material Crisaborole
CAS 906673-24-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H10BNO3
Quantity 100 mg
Synonym AN-2728 PF-06930164
Solubility Information DMSO : ≥ 100 mg/mL (398.33 mM)
SMILES N#CC1=CC=C(OC2=CC=C(B(O)OC3)C3=C2)C=C1
Molecular Weight (g/mol) 251.05
Formula Weight 251.05
Percent Purity 96.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.