missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISP-2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76277AF350
This item is not returnable.
View return policy
Description
CRISP-2 Polyclonal antibody specifically detects CRISP-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CRISP-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| Cancer/testis antigen 36, CRISP-2GAPDL5, CT36TSP1, cysteine-rich secretory protein 2, glyceraldehyde-3-phosphate dehydrogenase-like 5, testis specific protein 1 (probe H4-1 p3-1), Testis-specific protein TPX-1, TPX1MGC111136 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human CRISP2. The immunogen is located within the first 50 amino acids of CRISP2. Amino Acid Squence: CPSLPAEGKDPAFTAL | |
| 0.1 mL | |
| Signal Transduction | |
| 7180 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction