missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPLD2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97242AF488
This item is not returnable.
View return policy
Description
CRISPLD2 Polyclonal antibody specifically detects CRISPLD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CRISPLD2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83716 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| CRISP11, CRISP-11, Cysteine-rich secretory protein 11, cysteine-rich secretory protein LCCL domain containing 2, cysteine-rich secretory protein LCCL domain-containing 2, DKFZP434B044, LCCL domain containing cysteine-rich secretory protein 2, LCCL domain-containing cysteine-rich secretory protein 2, LCRISP2, MGC74865 | |
| Produced in rabbits immunized with E. coli-derived Human CRISPLD2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion