missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (6G12), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52398MFV450
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)
Specifications
| CRISPR-Cas9 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
| Western Blot, Immunocytochemistry, Immunoprecipitation, ChIP Assay | |
| 6G12 | |
| Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| This CRISPR-Cas9 antibody (6G12) - C-terminus was raised against recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2] | |
| 0.1 mL | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| 901176 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction