missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (7A9-3A3), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36440JF549
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Mouse, Bacteria, Bovine, Chicken, Fungi samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry (Whole-Mount), Gene Knock-Out
Specifications
| CRISPR-Cas9 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2]. | |
| 0.1 mL | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| 901176 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Simple Western, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 7A9-3A3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Bacteria, Bovine, Chicken, Fungi | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction