Learn More
Crizotinib, MedChemExpress
Product Code. 16494037
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494037 50 mg 50mg
16461926 500 mg 500mg
16461326 200 mg 200mg
16438128 10 mg 10mg
16437886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16494037 Supplier Medchem Express Supplier No. HY50878
MedChemExpress Crizotinib (PF-02341066) is an orally bioavailable, ATP-competitive ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively. Crizotinib inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 11 nM in cell-based assays, respectively. Crizotinib is also a ROS1 inhibitor. Crizotinib has effective tumor growth inhibition.
Crizotinib (PF-02341066) is an orally bioavailable, ATP-competitive ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively. Crizotinib inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 11 nM in cell-based assays, respectively. Crizotinib is also a ROS1 inhibitor. Crizotinib has effective tumor growth inhibition.
Specifications
Chemical Name or Material Crizotinib
CAS 877399-52-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H22Cl2FN5O
Quantity 50 mg
Synonym PF-02341066
Solubility Information DMSO : 33.33 mg/mL (74.01 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES ClC1=C(F)C=CC(Cl)=C1[C@H](OC2=CC(C3=CN(N=C3)C4CCNCC4)=CN=C2N)C
Molecular Weight (g/mol) 450.34
Formula Weight 450.34
Percent Purity 97.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only