Learn More
Crizotinib, Mikromol™
Description
Crizotinib
Specifications
Specifications
| Chemical Name or Material | Crizotinib |
| CAS | 877399-52-5 |
| Analyte or Component Names | Crizotinib |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21H22Cl2FN5O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-[(1R)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl)-1H-pyrazol-4-yl]-2-pyridinamine, (R)-3-[1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-(1-piperidin-4-yl-1H- pyrazol-4-yl)pyridin-2-ylamine, PF 02341066, PF 2341066, [3-[[(R)-1-(2,6-Dichloro-3-fluorophenyl)ethyl]oxy]-5-[1-(piperidin-4-yl)-1H-pyrazol-4-yl]pyridin-2-yl]amine |
| InChI Formula | InChI=1S/C21H22Cl2FN5O/c1-12(19-16(22)2-3-17(24)20(19)23)30-18-8-13(9-27-21(18)25)14-10-28-29(11-14)15-4-6-26-7-5-15/h2-3,8-12,15,26H,4-7H2,1H3,(H2,25,27)/t12-/m1/s1 |
| SMILES | CC(C1=C(C=CC(=C1Cl)F)Cl)OC2=C(N=CC(=C2)C3=CN(N=C3)C4CCNCC4)N |
| IUPAC Name | 3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-(1-piperidin-4-ylpyrazol-4-yl)pyridin-2-amine |
| Show More |