missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 3/PSA Antibody (rKLK3/6947) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20702
This item is not returnable.
View return policy
Description
Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kallikrein 3/PSA | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| Recombinant fragment (around aa150-250) of human Kallikrein 3/PSA protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers | |
| 354 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rKLK3/6947 | |
| Unconjugated | |
| 10mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction