missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CRLR Antibody (erenumab) - IgG2SA, Novus Biologicals™

Product Code. 30045780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30045780 100 μg 100µg
30047917 50 μg 50µg
30046476 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30045780 Supplier Novus Biologicals™ Supplier No. NBP328437100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CRLR Monoclonal antibody specifically detects CRLR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRLR
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone erenumab
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. Q16602
Gene Alias calcitonin gene-related peptide type 1 receptor, calcitonin receptor-like, CALCRL, CGRP type 1 receptor, CGRPR, CGRPRCRLRCalcitonin receptor-like receptor
Host Species Human
Immunogen CALCRL/ CGRPR
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10203
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.