missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76982MFV450
This item is not returnable.
View return policy
Description
CRMP1 Polyclonal antibody specifically detects CRMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CRMP1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| collapsin response mediator protein 1DRP1, CRMP-1, dihydropyrimidinase-like 1, dihydropyrimidinase-related protein 1, DPYSL1DRP-1dihydropyrimidinase related protein-1, ULIP3, ULIP-3, Unc-33-like phosphoprotein 3 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human CRMP1. The immunogen is located within amino acids 90 - 140 of CRMP1. Amino Acid Squence: SPGERDERPPTLRIRRPA | |
| 0.1 mL | |
| Neuroscience | |
| 1400 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction