missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68021-1-IG-20UL
This item is not returnable.
View return policy
Description
The CRMP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRMP1. This antibody recognizes mouse, rat, rabbit, pig, chicken, human antigen. The CRMP1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| CRMP1 | |
| Monoclonal | |
| 500 μg/mL | |
| DPYSL1; DRP-1; DRP1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Rabbit, Pig, Chicken, Human | |
| 62 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 2F4G5 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 1400 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction