missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97750-50ul
This item is not returnable.
View return policy
Description
CRMP2 Polyclonal antibody specifically detects CRMP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| CRMP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate | |
| Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
| Produced in rabbits immunized with E. coli-derived Human CRMP2 fragment. | |
| 50 μg | |
| Neuroscience | |
| 1808 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction