missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14521-1-AP-20UL
This item is not returnable.
View return policy
Description
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.Specifications
| CRMP2 | |
| Polyclonal | |
| Unconjugated | |
| DPYSL2 | |
| CRMP 2, CRMP2, DHPRP2, dihydropyrimidinase like 2, DPYSL2, DRP 2, DRP2, N2A3, ULIP 2, ULIP2, Unc 33 like phosphoprotein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12934, 1808, 25416 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08553, P47942, Q16555 | |
| DPYSL2 | |
| CRMP2 Fusion Protein Ag5987 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction