missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRMP2 Recombinant Rabbit Monoclonal Antibody (0A1T1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535510
This item is not returnable.
View return policy
Description
Immunogen sequence: PFPDFVYKRI KARSRLAELR GVPRGLYDGP VCEVSVTPKT VTPASSAKTS PAKQQAPPVR NLHQSGFSLS GAQIDDNIPR RTTQRIVAPP GGRANITSLG.
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
| CRMP2 | |
| Recombinant Monoclonal | |
| 0.233 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08553, P47942, Q16555 | |
| DPYSL2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0A1T1 | |
| Unconjugated | |
| DPYSL2 | |
| AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12934, 1808, 25416 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction