missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRMP5 Monoclonal Antibody (CR-3)
GREENER_CHOICE

Product Code. 11555782
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11555782 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11555782 Supplier Invitrogen™ Supplier No. MA3700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MA3-700 detects human, bovine, rat and mouse N-terminal region of CRMP5. MA3-700 has been successfully used in Western blot and immunohistochemical procedures. By Western blot, this antibody detects an ~ 62 kDa protein corresponding to human CRMP5 from brain extract. The MA3-700 antigen is a purified protein from human brain. This antibody is specific for the N-terminal region, amino acids 1-64. Not reactive with other CRMP isoforms.

The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP5
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CR-3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene DPYSL5
Gene Accession No. Q9BPU6, Q9EQF6, Q9JHU0
Gene Alias collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6
Gene Symbols DPYSL5
Host Species Rat
Immunogen CRMP5 protein purified from human brain.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100126171, 56896, 65208, 65254
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.