missing translation for 'onlineSavingsMsg'
Learn More

Crocein Orange G, TRC

Product Code. 30362551
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362551 500 mg -
30352073 250 mg 250mg
30358430 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362551 Supplier Toronto Research Chemicals Supplier No. TRCC794955500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-20-9
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.32
Synonym Brilliant Orange (6CI), C.I. Acid Orange 12 (7CI), C.I. Acid Orange 12, monosodium salt (8CI), 1008 Orange, Acid Bright Orange Zh, Acid Brilliant Orange Zh, Acid Orange 12, Acidine Orange GN, Acilan Orange G, Acilan Ponceau 4GBLAmacid Brilliant Orange, Basic Brilliant orange G, Brilliant Orange G, Brilliant Orange GN, Brilliant Orange GN Type 8019, C.I. 15970, C.I. Food Orange 1, Crocein Orange, Crocein Orange G, Croceine Orange, Croceine Orange 2G, Croceine Orange EN, Croceine Orange G, Croceine Orange Y, Food orange No. 1, Helio Orange CAG, Hexacol Orange RN, Hispacid Orange CG, Kiton Brilliant Orange G, Kiton Ponceau 4G, Lutetia Orange 2JR, Monolite Orange C, Orange G Food Grade, Orange LZS, Orange RN, Ponceau 4G, Segnale Light Orange GR, Siloton Orange GR, Sodium 1-(Phenylazo)-2-naphthol-6-sulfonate, Sodium 6-Hydroxy-5-(phenylazo)-2-naphthalenesulfonate, Tertracid Brilliant Orange P 4G, 6-Hydroxy-5-(phenylazo)-2-naphthalenesulfonic Acid Monosodium Salt (9CI)
IUPAC Name sodium;6-hydroxy-5-[(E)-phenyldiazenyl]naphthalene-2-sulfonate
SMILES [Na+].Oc1ccc2cc(ccc2c1N=Nc3ccccc3)S(=O)(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H12N2O4S.Na/c19-15-9-6-11-10-13(23(20,21)22)7-8-14(11)16(15)18-17-12-4-2-1-3-5-12;/h1-10,19H,(H,20,21,22);/q;+1/p-1
Formula Weight 350.0337
Chemical Name or Material Crocein Orange G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.