missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CROT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553984
This item is not returnable.
View return policy
Description
Immunogen sequence: VRWGDKSYNL ISFSNGVFGC NCDHAPFDAM IMVNISYYVD EKIFQNEGRW KGSEKVRDIP LPEELIFIVD EKVLNDINQA KAQYLR Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
Specifications
| CROT | |
| Polyclonal | |
| Unconjugated | |
| Crot | |
| 1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKG9 | |
| Crot | |
| Recombinant protein corresponding to Human CROT. Recombinant protein control fragment (Product #RP-92591). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction