Learn More
Crotonaldehyde 2,4-Dinitrophenylhydrazone-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Crotonaldehyde 2,4-Dinitrophenylhydrazone-d3 |
| CAS | 259824-64-1 |
| Molecular Formula | C10 D3 H7 N4 O4 |
| Quantity | 50 mg |
| Synonym | 2-Butenal, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9CI), 2-Butenal (2,4-dinitrophenyl-3,5,6-d3)hydrazone, Crotonaldehyde-2,4-dinitrophenylhydrazone-d3, Crotonaldehyde-2,4-dinitrophenylhydrazone-D3 (phenyl-D3) |
| InChI Formula | InChI=1S/C10H10N4O4/c1-2-3-6-11-12-9-5-4-8(13(15)16)7-10(9)14(17)18/h2-7,12H,1H3/b3-2+,11-6+/i4D,5D,7D |
| SMILES | [2H]c1c([2H])c(c([2H])c(c1N\N=C\C=C\C)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | N-[(E)-[(E)-but-2-enylidene]amino]-2,3,5-trideuterio-4,6-dinitroaniline |
| Molecular Weight (g/mol) | 253.229 |
| Formula Weight | 253.089 |