missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Crotonylated Lysine Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5120739

Product Code. 17772134

  • € 514.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive test controls include: HeLa, NIH/3T3, Mouse brain, Mouse testis, Rat brain, Rat testis.

A Post translational modification to the amino acid lysine. It is being studied for links to diseased states.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Crotonylated Lysine
Polyclonal
Unconjugated
Kcr; Lysine
A synthetic peptide corresponding to a sequence containing crotonylated K.
100 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, Western Blot
0.44 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Rabbit
Affinity Chromatography
RUO
Chemical, Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.