Learn More
LRRC32/GARP Antibody (725226), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB62291PEATT594
Product Code. 30023630
  • ONLINE EXCLUSIVE PRICE
    € 674.00
    € 637.35 / 0.10mL
    Save € 36.65
    5% Off
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRRC32/GARP
Monoclonal
PE/ATTO 594
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
2615
Store at 4C in the dark. Do not freeze.
IgG1
Flow Cytometry
725226
Flow Cytometry
D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP, Ile18-Asn628, Accession # NP_001106850
0.1 mL