missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH-2/GPR44 Antibody (301108) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3338C
This item is not returnable.
View return policy
Description
CRTH-2/GPR44 Monoclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CRTH-2/GPR44 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 301108 | |
| Flow Cytometry, CyTOF-ready | |
| CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
| NS0 mouse myeloma cell line transfected with human CRTH-2, Met1-Ser395, Accession # Q9Y5Y4 | |
| 0.1 mL | |
| GPCR, Innate Immunity | |
| 11251 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction