missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH-2/GPR44 Antibody (54N1H4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-19013AF405
This item is not returnable.
View return policy
Description
CRTH-2/GPR44 Monoclonal specifically detects CRTH-2/GPR44 in Human samples. It is validated for Western Blot.
Specifications
| CRTH-2/GPR44 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| GPR44, or G-protein coupled receptor 44, is also designated as CRTH2 and CD294. GPR44 is expressed specifically on CD4+ Th2 cells and type 2 cytotoxic effector cells | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot | |
| 54N1H4 | |
| Western Blot | |
| PTGDR2 | |
| A recombinant protein containing multiple external domains of GPR44 (CRTH2) was used as immunogen for the antibody. NP_004769 | |
| 0.1 mL | |
| GPCR | |
| 11251 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction