missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH-2/GPR44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-02355
This item is not returnable.
View return policy
Description
CRTH-2/GPR44 Polyclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CRTH-2/GPR44 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
| Synthetic 20 amino acid peptide from C-terminal cytoplasmic domain of human CRTH-2/GPR44. | |
| 0.05 mg | |
| GPCR, Innate Immunity | |
| 11251 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry 15 μg/mL, Immunohistochemistry-Paraffin 15 μg/mL | |
| Q9Y5Y4 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction