Learn More
MedChemExpress Crustacean Cardioactive Peptide (CCAP)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Purity | 99.66% |
| CAS | 309247-84-5 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 959.1 g/mol |
| Product Type | Natural Products |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 50 mg/mL (52.13 mM∣Need ultrasonic) ∣H2O : 0.67 mg/mL (0.70 mM∣Need ultrasonic) |