missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRX/CORD2 Antibody (SR1615), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21935-50ul
This item is not returnable.
View return policy
Description
CRX/CORD2 Monoclonal antibody specifically detects CRX/CORD2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CRX/CORD2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1406 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| SR1615 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| cone-rod homeobox, CORD2, CRDcone-rod homeobox protein, LCA7orthodenticle homeobox 3, OTX3 | |
| A synthesized peptide derived from human CRX/CORD2 (Uniprot #: O43186) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction