missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CRX/CORD2 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Packungsgröße:
0.1mL
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CRX. The CRX/CORD2 Recombinant Protein Antigen is derived from E. coli. The CRX/CORD2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-88059. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Spezifikation
Spezifikation
| Gene ID (Entrez) | 1406 |
| Species | Human |
| Purification Method | Chromatography |
| Purity | >80% |
| Concentration | 0.5mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Symbol | CRX |
| Label Type | Unlabeled |
| Mehr anzeigen |
For Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur