missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ cry2Ab Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144423
This item is not returnable.
View return policy
Description
cry2Ab Polyclonal Antibody for Western Blot, ELISA
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of lepidopteran (Manduca sexta) larvae.
Specifications
| cry2Ab | |
| Polyclonal | |
| Unconjugated | |
| cryB2; cryIIA | |
| Recombinant Bacillus thuringiensis subsp.Kurstaki Pesticidal crystal protein cry2Ab (cry2Ab) (1-633aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.06 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction