missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRYAB Recombinant Rabbit Monoclonal Antibody (K01_1M64)

Product Code. 30828168
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828168 50 μL
30828472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828168 Supplier Invitrogen™ Supplier No. MA564593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: TREEKPAVTA APKK.

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYAB
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1M64
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Cryab
Gene Accession No. P02511, P23928
Gene Alias 2310001E10Rik; 3526402H21Rik; AACRYA; active transcription factor CREB; alpha B-crystallin; alpha crystallin B; alpha(B)-crystallin; alpha-B-crystallin; Alpha-crystallin B chain; alpha-crystallin B chain-like protein; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CMD1II; CREB; CREB1; CREB-1; Crya2; Crya-2; CRYAB; CryAB antibody; crystallin alpha B; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; CTPP2; CTRCT16; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; epididymis secretory protein Li 101; heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HEL-S-101; HSPB5; HspB5 protein; MFM2; P23; Renal carcinoma antigen NY-REN-27; rosenthal fiber component; transactivator protein; Y protein
Gene Symbols Cryab
Host Species Rabbit
Immunogen A synthetic peptide of human Alpha B Crystallin (aa 162-175).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410, 25420
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.