missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRYBB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16399054
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16399054 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16399054 Supplier Invitrogen™ Supplier No. PA582093

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TTVPITSAKA AELPPGNYRL VVFELENFQG RRAEFSGECS NLADRGFDRV RSIIVSAGPW VAFEQSNFRG EMFILEKGEY PRWNTWSSSY RSDRLMSFRP IKMDAQE Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 80%.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta basic group member, undergoes extensive cleavage at its N-terminal extension during lens maturation. It is also a member of a gene cluster with beta-A4, beta-B2, and beta-B3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYBB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Crybb1
Gene Accession No. P53674
Gene Alias 3110006K12Rik; BB1CRY; Beta-B1 crystallin; beta-crystallin B1; Beta-crystallin B1B; CATCN3; CRYB1; CRYB11; Crybb1; crystallin beta B1; crystallin, beta B1; CTRCT17; eye lens structural protein
Gene Symbols Crybb1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRYBB1. Recombinant protein control fragment (Product #RP-108304).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.