missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYGA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570666
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 79%; Zebrafish: 82%.
CRYGA is a protein coding gene.
Specifications
| CRYGA | |
| Polyclonal | |
| Unconjugated | |
| CRYGA | |
| CRYG1; Cryg-4; CRYG5; CRYGA; CRY-g-A; CRY-gamma-A; crystallin gamma A; crystallin, gamma 1; crystallin, gamma A; crystallin, gamma polypeptide 1; DGcry-4; Gamma-A-crystallin; gammaA-crystallin; Gamma-crystallin 1-1; gamma-crystallin 4; Gamma-crystallin 5; gamma-crystallin A; Gamma-crystallin IVB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1418 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P11844 | |
| CRYGA | |
| synthetic peptide directed towards the N-terminal region of human CRYGA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction