Learn More
SPEX Certiprep™ Cryogenic Grinding Vial Pack
Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
SPEX Homogenizer
Unit Size:
Pack of 4
Description
- A complete vial consists of 2 end plugs, a polycarbonate cylinder and 2 stainless steel 1/2” (12.7 mm) grinding balls.
- Vials are sold in sets of 4.
- Up to 16 vials can be run simultaneously
- Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
Specifications
Specifications
| For Use With (Application) | Use with the SPEX homogeniser 2260 CryoBlock |
| For Use With (Equipment) | SPEX Homogenizer |
| Material | Polycarbonate |
| Packaging Quantity | 4 Set |