Learn More
CellPath Cryospray Freezer Spray
Description
- CFC free
- Directable jet enables precise control
- Ideal for paraffin blocks and fresh tissue specimens
- Manufactured from a mixture of environmentally friendly gases to give a good balance of cooling efficiency and cost
- Supplied in a compact seamless aluminum can that is easy to handle, with a nozzle that ensures accurate direction of the freezing jet
Specifications
Specifications
| Capacity (Metric) | 175 mg |
| Type | Cryospray Freezer Spray |
| No. per Pack | 1 Pack |
| For Use With (Application) | Ideal for paraffin blocks and fresh tissue specimens |
| Quantity | 175 mg |
Safety and Handling
Product Identifier
- CRYOSPRAY (FREEZER SPRAY)
- Warning
- Flammable and non-flammable aerosols category 1
- H229-Pressurised container: May burst if heated.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P251-Do not pierce or burn, even after use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P410-Protect from sunlight.
- P412-Do not expose to temperatures exceeding 50°C/122°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 1,1,1,2-Tetrafluoroethane