Learn More
STEMCELL Technologies CryoStor™ CS10
Product Code. 17159931
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL Bag
1000 mL Bag
5 x 10 mL
5 x 16 mL
Unit Size:
1000mL
100mL
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159931 100 mL Bag 100mL
15642424 5 x 16 mL Pack of 5
17169931 5 x 10 mL Pack of 5
17199921 1000 mL Bag 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17159931 Supplier STEMCELL Technologies Supplier No. 07955
Animal-component-free, defined cryopreservation medium with 10% DMSO
  • CryoStor™ CS10 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS10 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS10 is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, CHO cells, myeloma cell lines, hybridomas, human mesenchymal stem cells, human embryonic and human induced pluripotent stem cells (ES cells and iPS cells) and other extremely sensitive cell types
  • CryoStor™ CS10 is cGMP-manufactured with USP grade components
Specifications
Cell Type B Cells, CHO Cells, Hematopoietic Stem and Progenitor Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells
For Use With (Application) Cryopreservation
Quantity 100 mL Bag