missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cryptic Monoclonal Antibody (221423)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561051
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| Cryptic | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P97766 | |
| CFC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cryptic, Tyr36-Gly177. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 221423 | |
| Unconjugated | |
| CFC1 | |
| AV265756; b2b970Clo; Cfc1; CFC-1; CFC1B; cripto; cripto, FRL-1, cryptic family 1; cryptic; cryptic family 1; Cryptic family protein 1; cryptic protein; CRYPTICFLJ77897; DTGA2; heterotaxy 2 (autosomal dominant); HTX2; HTX2MGC133213 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 12627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction