missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium Antibody (BEL 0126), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64526APCCY7
This item is not returnable.
View return policy
Description
Cryptosporidium Monoclonal specifically detects Cryptosporidium in Protozoa samples. It is validated for Flow Cytometry.
Specifications
| Cryptosporidium | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Protozoa | |
| IgG3 |
| Flow Cytometry | |
| BEL 0126 | |
| Flow Cytometry | |
| Cryptosporidiosis | |
| Purified oocysts from bovine faeces | |
| 0.1 mL | |
| Recognizes a membrane antigen expressed by the oocysts of Cryptosporidium sp., an obligate intracellular parasite of the phylum Apicomplexa. The genome of Cryptosporidium sp. is one of the smallest among unicellular eukaryotic organisms. Cryptosporidium is one of several species that cause cryptosporidiosis; a disease affecting the intestines of mammals which is spread through the faecal-oral route. The main symptom is self-limiting diarrhoea in people with intact immune systems. However, in immunocompromised individuals, such as AIDS patients, infection can cause permanent & life-threatening diarrhoea. Cryptosporidiosis is one of the most common waterborne diseases worldwide. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction