missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium Antibody (BEL 0126), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64526F
This item is not returnable.
View return policy
Description
Cryptosporidium Monoclonal antibody specifically detects Cryptosporidium in Protozoa samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Cryptosporidium | |
| Monoclonal | |
| FITC | |
| Cryptosporidiosis | |
| Purified oocysts from bovine faeces | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| BEL 0126 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction