missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium Parvum Antibody (7631), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65674B
This item is not returnable.
View return policy
Description
Cryptosporidium Parvum Monoclonal specifically detects Cryptosporidium Parvum in Mouse, Protozoa samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cryptosporidium Parvum | |
| Monoclonal | |
| 0.1mg/mL | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-65674 detects the protozoa Cryptosporidium parvum, an obligate intracellular parasite of the phylum Apicomplexa. The genome of C. parvum is one of the smallest among unicellular eukaryotic organisms. C. parvum is one of several species that cause cryptosporidiosis; a disease affecting the intestines of mammals and is spread through the fecal-oral route. The main symptom is self-limiting diarrhea in people with intact immune systems. However, in immunocompromised individuals, such as AIDS patients, infection can cause permanent & life-threatening diarrhea. Cryptosporidiosis is one of the most common waterborne diseases worldwide. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7631 | |
| Biotin | |
| C. parvum | |
| Intact oocysts of C.Parvum. | |
| 0.05 mL | |
| Primary | |
| Mouse, Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction