missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium Parvum Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-1479
This item is not returnable.
View return policy
Description
Cryptosporidium Parvum Polyclonal specifically detects Cryptosporidium Parvum in Protozoa samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cryptosporidium Parvum | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (pH 7.2) with 0.1% Sodium Azide | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| This is specific to intact Oocysts. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 4.5 mg/mL | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| C. parvum | |
| Oocysts purified from bovine feces. | |
| 0.1 mL | |
| Primary | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction