Learn More
Invitrogen™ Cryptosporidium parvum Oocysts Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173186
Description
The PA1-73186 antibody reacts with intact Cryptosporidium parvum oocysts. PA1-73186 has been successfully used in ELISA and Immunofluorescence applications. The PA1-73186 antibody was raised against oocysts purified from bovine feces. Recommended short term storage of less than 6 months at 2-8°C. For extended storage, aliquot and freeze.
Specifications
| Cryptosporidium parvum Oocysts | |
| Polyclonal | |
| Biotin | |
| Cryptosporidiosis | |
| Oocysts purified from bovine feces. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Protozoa | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.