Learn More
Multiroir Controlec™ Crystal ABS Storage Box
Description
- Rugged and shock resistant
- Completely transparent
- Lid included
Specifications
Specifications
| Color | White |
| Material | ABS |
| Shape | Rectangular |
| Product Type | Tilt Bin |
| Length (Metric) | 122 mm |
| Width (Metric) | 133 mm |
| Height (Metric) | 160 mm |
| Weight (Metric) | 0.47 kg |